siRNA Details
| Gene Symbol: LOC690352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109582.1 | NP_001103052.1 | 6 | 742 | ||
| XM_006251549.2 | XP_006251611.1 | 5 | 729 | |||
| XM_006251550.2 | XP_006251612.1 | 5 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690352| Gene Symbol: LOC690352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109582.1 | NP_001103052.1 | 6 | 742 | ||
| XM_006251549.2 | XP_006251611.1 | 5 | 729 | |||
| XM_006251550.2 | XP_006251612.1 | 5 | 626 | |||