siRNA Details
| Gene Symbol: Cnih3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166578.1 | NP_001160050.1 | 1, 2 | 88 | ||
| GenBank mRNA | FJ403327.1 | - | 1, 2 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnih3| Gene Symbol: Cnih3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166578.1 | NP_001160050.1 | 1, 2 | 88 | ||
| GenBank mRNA | FJ403327.1 | - | 1, 2 | 88 | ||