siRNA Details
Gene Symbol: Muc5ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760037.1 | XP_008758259.1 | 28 | 3757 | ||
GenBank mRNA | AB042530.1 | - | 28 | 3784 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Muc5ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760037.1 | XP_008758259.1 | 28 | 3757 | ||
GenBank mRNA | AB042530.1 | - | 28 | 3784 |