siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134642.1 | NP_001128114.1 | 8 | 983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem194b| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134642.1 | NP_001128114.1 | 8 | 983 | ||