siRNA Details
| Gene Symbol: Apcdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071384.4 | XP_001071384.2 | 2 | 193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apcdd1| Gene Symbol: Apcdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001071384.4 | XP_001071384.2 | 2 | 193 | ||