siRNA Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107115.1 | NP_001100585.1 | 2 | 344 | ||
| XM_006248760.2 | XP_006248822.1 | 2 | 121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef18| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107115.1 | NP_001100585.1 | 2 | 344 | ||
| XM_006248760.2 | XP_006248822.1 | 2 | 121 | |||