siRNA Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107115.1 | NP_001100585.1 | 4 | 609 | ||
| XM_006248760.2 | XP_006248822.1 | 5 | 633 | |||
| XM_008768950.1 | XP_008767172.1 | 3 | 191 | |||
| XM_008768952.1 | XP_008767174.1 | 3 | 191 | |||

Arhgef18
Genomic Map