siRNA Details
| Gene Symbol: Lyrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126096.1 | NP_001119568.1 | 2 | 107 | ||
| GenBank mRNA | BC166578.1 | - | 2 | 107 | 7379886 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyrm2| Gene Symbol: Lyrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126096.1 | NP_001119568.1 | 2 | 107 | ||
| GenBank mRNA | BC166578.1 | - | 2 | 107 | 7379886 | |