siRNA Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128189.2 | NP_001121661.1 | 10, 11 | 1253 | ||
| XM_006239073.2 | XP_006239135.1 | 10, 11 | 1223 | |||
| XM_006239074.1 | XP_006239136.1 | 10, 11 | 1547 | |||
| XM_006239075.2 | XP_006239137.1 | 10, 11 | 1737 | |||
| XM_006239077.2 | XP_006239139.1 | 9, 10 | 1183 | |||
| XM_008764172.1 | XP_008762394.1 | 10, 11 | 2891 | |||
| XM_008764173.1 | XP_008762395.1 | 11, 12 | 1277 | |||
| GenBank mRNA | BC160834.1 | - | 10, 11 | 1253 | 7456254 | |

Rcc1
Genomic Map