siRNA Details
| Gene Symbol: Cntnap5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047865.1 | NP_001041330.1 | 3373 | |||
| Gene Symbol: LOC100910512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769395.1 | XP_008767617.1 | 1218 | |||

Cntnap5a
Genomic Map