siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769959.1 | XP_008768181.1 | 5 | 1059 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_018759, AK146438, AK017693, BC005567, AB012725.

Zfp326
Genomic Map