siRNA Details
| Gene Symbol: Atp8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751492.3 | XP_003751540.1 | 4 | 2064 | ||
| XM_006252160.2 | XP_006252222.1 | 4 | 731 | |||
| XM_006252164.2 | XP_006252226.1 | 4 | 1602 | |||
| XM_006252165.2 | XP_006252227.1 | 4 | 336 | |||
| XM_008770789.1 | XP_008769011.1 | 4 | 1475 | |||
| XM_008770790.1 | XP_008769012.1 | 4 | 792 | |||
| XM_008770791.1 | XP_008769013.1 | 5 | 460 | |||

Atp8a2
Genomic Map