siRNA Details
| Gene Symbol: LOC102549842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227817.2 | XP_006227879.1 | 4 | 3140 | ||
| XM_008758701.1 | XP_008756923.1 | 4 | 3989 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102549842| Gene Symbol: LOC102549842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227817.2 | XP_006227879.1 | 4 | 3140 | ||
| XM_008758701.1 | XP_008756923.1 | 4 | 3989 | |||