siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144840.1 | NP_001138312.1 | 12 | 1717 | ||
| XM_006248776.2 | XP_006248838.1 | 13 | 1824 | |||
| XM_006248777.2 | XP_006248839.1 | 13 | 1821 | |||
| XM_006248778.2 | XP_006248840.1 | 12 | 1791 | |||
| XM_006248779.2 | XP_006248841.1 | 12 | 1776 | |||
| XM_006248781.2 | XP_006248843.1 | 11 | 1743 | |||
| XM_006248782.2 | XP_006248844.1 | 11 | 1740 | |||
| XM_008769001.1 | XP_008767223.1 | 12 | 1773 | |||

Camsap3
Genomic Map