siRNA Details
| Gene Symbol: Rnf11l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271224.1 | NP_001258153.1 | 3 | 794 | ||
| Gene Symbol: Rnf11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245310.2 | XP_006245372.1 | 1 | 621 | ||

Rnf11l1
Genomic Map