siRNA Details
| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014641.2 | NP_055456.2 | 5 | 2356 | ||
| XM_005249492.2 | XP_005249549.1 | 5 | 2260 | |||
| XM_005249493.3 | XP_005249550.1 | 5 | 2260 | |||
| XM_005249494.4 | XP_005249551.1 | 5 | 2659 | |||
| XM_005249497.3 | XP_005249554.1 | 5 | 2260 | |||
| XM_005249498.3 | XP_005249555.1 | 5 | 2260 | |||
| XM_011515001.2 | XP_011513303.1 | 5 | 2087 | |||
| XM_011515003.2 | XP_011513305.1 | 5 | 1942 | |||
| XM_011515004.2 | XP_011513306.1 | 6 | 1971 | |||
| XM_017011519.1 | XP_016867008.1 | 5 | 2039 | |||
| XM_017011520.1 | XP_016867009.1 | 5 | 1958 | |||
| XM_017011521.1 | XP_016867010.1 | 5 | 1958 | |||
| XM_017011522.1 | XP_016867011.1 | 5 | 2260 | |||
| GenBank mRNA | AK302434.1 | - | 6 | 1962 | ||
| BC110645.1 | - | 1 | 1082 | |||
| BC152556.1 | - | 4 | 1929 | 9007258 | ||
| D79992.1 | - | 4 | 1929 | |||

MDC1
Genomic Map