siRNA Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752031.3 | XP_003752079.2 | 3 | 472 | ||
| XM_006256807.2 | XP_006256869.1 | 3 | 5239 | |||
| XM_006256808.2 | XP_006256870.1 | 3 | 1325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem29| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752031.3 | XP_003752079.2 | 3 | 472 | ||
| XM_006256807.2 | XP_006256869.1 | 3 | 5239 | |||
| XM_006256808.2 | XP_006256870.1 | 3 | 1325 | |||