siRNA Details
| Gene Symbol: ZNF516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014643.3 | NP_055458.1 | 4 | 3511 | ||
| XM_011526269.1 | XP_011524571.1 | 3 | 3449 | |||
| XM_011526270.2 | XP_011524572.1 | 3 | 3712 | |||
| XM_011526271.2 | XP_011524573.1 | 4 | 3826 | |||
| XM_011526272.2 | XP_011524574.1 | 4 | 3547 | |||
| XM_011526273.2 | XP_011524575.1 | 3 | 3462 | |||
| XM_011526274.2 | XP_011524576.1 | 3 | 3468 | |||
| XM_011526275.2 | XP_011524577.1 | 3 | 4325 | |||
| XM_011526276.1 | XP_011524578.1 | 3 | 3449 | |||
| XM_011526277.2 | XP_011524579.1 | 3 | 3450 | |||
| XM_017026097.1 | XP_016881586.1 | 4 | 3582 | |||
| GenBank mRNA | BC108287.1 | - | 1 | 1365 | ||
| D86975.1 | - | 4 | 3511 | |||

ZNF516
Genomic Map