siRNA Details
| Gene Symbol: C1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106021.1 | NP_001099491.1 | 5 | 381 | ||
| XM_006251516.2 | XP_006251578.1 | 6 | 500 | |||
| XM_008770422.1 | XP_008768644.1 | 5 | 2563 | |||
| XM_008770423.1 | XP_008768645.1 | 6 | 2462 | |||
| GenBank mRNA | BC168684.1 | - | 5 | 357 | 7930158 | |
| FQ215730.1 | - | 5 | 355 | |||
| FQ229879.1 | - | 5 | 310 | |||
| FQ229896.1 | - | 5 | 359 | |||

C1d
Genomic Map