siRNA Details
| Gene Symbol: CEP135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025009.4 | NP_079285.2 | 5 | 818 | ||
| XM_005265788.3 | XP_005265845.1 | 5 | 789 | |||
| XM_006714055.2 | XP_006714118.1 | 5 | 818 | |||
| GenBank mRNA | AK023683.1 | - | 5 | 755 | ||
| AK309554.1 | - | 2 | 155 | |||
| BC136535.1 | - | 5 | 645 | 9020525 | ||
| BC136536.1 | - | 5 | 645 | 9020526 | ||

CEP135
Genomic Map