siRNA Details
| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079711.1 | NP_001073179.1 | 5, 6 | 596 | ||
| GenBank mRNA | BC128747.1 | - | 5, 6 | 596 | 7099220 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad51ap1| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079711.1 | NP_001073179.1 | 5, 6 | 596 | ||
| GenBank mRNA | BC128747.1 | - | 5, 6 | 596 | 7099220 | |