siRNA Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122781.3 | NP_001116253.3 | 5 | 610 | ||
| XM_006242354.2 | XP_006242416.1 | 5 | 673 | |||
| GenBank mRNA | BC159426.1 | - | 5 | 591 | 9034024 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Letmd1| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122781.3 | NP_001116253.3 | 5 | 610 | ||
| XM_006242354.2 | XP_006242416.1 | 5 | 673 | |||
| GenBank mRNA | BC159426.1 | - | 5 | 591 | 9034024 | |