siRNA Details
| Gene Symbol: Gdpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229796.2 | XP_006229858.1 | 12 | 1384 | ||
| XM_006229799.2 | XP_006229861.1 | 12 | 1382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdpd4| Gene Symbol: Gdpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229796.2 | XP_006229858.1 | 12 | 1384 | ||
| XM_006229799.2 | XP_006229861.1 | 12 | 1382 | |||