siRNA Details
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759584.1 | XP_008757806.1 | 31 | 3391 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fanci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759584.1 | XP_008757806.1 | 31 | 3391 |