siRNA Details
| Gene Symbol: LOC100363064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769049.1 | XP_008767271.1 | 2, 3 | 207 | ||
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068161.5 | XP_001068161.4 | 2, 3 | 189 | ||

LOC100363064
Genomic Map