siRNA Details
| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007741.1 | NP_001007742.1 | 2 | 849 | ||
| GenBank mRNA | BC079314.1 | - | 2 | 849 | 7134903 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isg20l2| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007741.1 | NP_001007742.1 | 2 | 849 | ||
| GenBank mRNA | BC079314.1 | - | 2 | 849 | 7134903 | |