siRNA Details
Gene Symbol: Trnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069068.5 | XP_001069068.5 | 1 | 1136 | ||
XM_008764186.1 | XP_008762408.1 | 1 | 1136 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069068.5 | XP_001069068.5 | 1 | 1136 | ||
XM_008764186.1 | XP_008762408.1 | 1 | 1136 |