siRNA Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322284.1 | NP_001309213.1 | 5 | 1384 | ||
| NM_001322285.1 | NP_001309214.1 | 5 | 1373 | |||
| NM_014650.3 | NP_055465.1 | 5 | 1782 | |||
| GenBank mRNA | AB018341.1 | - | 5 | 1373 | ||
| AK292827.1 | - | 5 | 1515 | |||
| BC002858.2 | - | 5 | 1478 | 3636865 | ||

ZNF432
Genomic Map