siRNA Details
| Gene Symbol: Chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762448.1 | XP_008760670.1 | 3 | 852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp4b| Gene Symbol: Chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762448.1 | XP_008760670.1 | 3 | 852 | ||