siRNA Details
| Gene Symbol: Plekhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190999.1 | NP_001177928.1 | 14 | 1947 | ||
| XM_006227838.2 | XP_006227900.1 | 15 | 5351 | |||
| XM_006227841.2 | XP_006227903.1 | 16 | 2282 | |||
| XM_006227842.2 | XP_006227904.1 | 16 | 2300 | |||
| XM_006227844.2 | XP_006227906.1 | 15 | 3076 | |||
| XM_008758706.1 | XP_008756928.1 | 15 | 2191 | |||
| XM_008758707.1 | XP_008756929.1 | 15 | 2194 | |||

Plekhg1
Genomic Map