siRNA Details
| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728846.4 | XP_002728892.3 | 7 | 2971 | ||
| XM_006230915.2 | XP_006230977.1 | 6 | 2896 | |||
| XM_006230916.2 | XP_006230978.1 | 6 | 2884 | |||
| GenBank mRNA | BC088266.1 | - | 4 | 262 | ||
| BC158837.1 | - | 5 | 436 | |||

Phemx
Genomic Map