siRNA Details
| Gene Symbol: LOC683313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101007.1 | NP_001094477.1 | 8 | 2095 | ||
| GenBank mRNA | BC099121.1 | - | 8 | 2095 | 7462749 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC683313| Gene Symbol: LOC683313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101007.1 | NP_001094477.1 | 8 | 2095 | ||
| GenBank mRNA | BC099121.1 | - | 8 | 2095 | 7462749 | |