siRNA Details
| Gene Symbol: Zik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228165.2 | XP_006228227.1 | 4 | 1152 | ||
| Gene Symbol: LOC100911224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228161.2 | XP_006228223.1 | 4 | 1025 | ||

Zik1
Genomic Map