siRNA Details
| Gene Symbol: Gtpbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771688.1 | XP_008769910.1 | 2 | 175 | ||
| GenBank mRNA | AF325355.1 | - | 2 | 100 | ||
| Gene Symbol: LOC299312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773045.1 | XP_008771267.1 | 1 | 128 | ||

Gtpbp4
Genomic Map