siRNA Details
| Gene Symbol: Gtpbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771688.1 | XP_008769910.1 | 7, 8 | 929 | ||
| GenBank mRNA | AF325355.1 | - | 7, 8 | 854 | ||
| CD371777.1 | - | 4, 5 | 484 | |||
| Gene Symbol: LOC299312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773045.1 | XP_008771267.1 | 1 | 873 | ||

Gtpbp4
Genomic Map