siRNA Details
| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060013.5 | XP_001060013.1 | 10, 11 | 1019 | ||
| XM_008766177.1 | XP_008764399.1 | 9, 10 | 996 | |||
| XM_008766178.1 | XP_008764400.1 | 10, 11 | 1040 | |||

Ccdc84
Genomic Map