siRNA Details
| Gene Symbol: Lilrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100123.1 | NP_001093593.1 | 3 | 375 | ||
| XM_006228169.2 | XP_006228231.1 | 2 | 422 | |||
| XM_006228170.2 | XP_006228232.1 | 3 | 460 | |||
| GenBank mRNA | AY998483.1 | - | 3 | 354 | ||

Lilrc2
Genomic Map