siRNA Details
| Gene Symbol: WSCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304447.1 | NP_001291376.1 | 9 | 2030 | ||
| NM_014653.3 | NP_055468.2 | 8 | 2199 | |||
| XM_017020243.1 | XP_016875732.1 | 9 | 2484 | |||
| XM_017020244.1 | XP_016875733.1 | 10 | 2645 | |||
| XM_017020245.1 | XP_016875734.1 | 10 | 2142 | |||
| XM_017020246.1 | XP_016875735.1 | 8 | 1873 | |||
| XM_017020247.1 | XP_016875736.1 | 9 | 2016 | |||
| XM_017020248.1 | XP_016875737.1 | 9 | 2488 | |||
| XM_017020249.1 | XP_016875738.1 | 8 | 2345 | |||
| XM_017020250.1 | XP_016875739.1 | 6 | 1899 | |||
| XM_017020251.1 | XP_016875740.1 | 7 | 1710 | |||
| XM_017020252.1 | XP_016875741.1 | 7 | 2188 | |||
| GenBank mRNA | AB018332.2 | - | 8 | 1936 | ||
| AK293764.1 | - | 7 | 1458 | |||
| BC037969.1 | - | 3 | 425 | |||
| BC110330.1 | - | 8 | 1982 | 5735001 | ||
| BC136687.1 | - | 7 | 1728 | 9020677 | ||
| BC136688.1 | - | 7 | 1728 | 9020678 | ||
| BX647283.1 | - | 9 | 1981 | |||

WSCD2
Genomic Map