siRNA Details
| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109529.1 | NP_001102999.1 | 5 | 1071 | ||
| XM_006254252.2 | XP_006254314.1 | 5 | 1251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kin| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109529.1 | NP_001102999.1 | 5 | 1071 | ||
| XM_006254252.2 | XP_006254314.1 | 5 | 1251 | |||