siRNA Details
| Gene Symbol: LOC683410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765862.1 | XP_008764084.1 | 4 | 1065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC683410| Gene Symbol: LOC683410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765862.1 | XP_008764084.1 | 4 | 1065 | ||