siRNA Details
| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771897.1 | XP_008770119.1 | 3 | 584 | ||
| XM_008771898.1 | XP_008770120.1 | 3 | 812 | |||
| XM_008771899.1 | XP_008770121.1 | 3 | 581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Proser2| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771897.1 | XP_008770119.1 | 3 | 584 | ||
| XM_008771898.1 | XP_008770120.1 | 3 | 812 | |||
| XM_008771899.1 | XP_008770121.1 | 3 | 581 | |||