siRNA Details
Gene Symbol: LOC688765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109512.1 | NP_001102982.1 | 1 | 562 | ||
XM_006253209.2 | XP_006253271.1 | 1 | 905 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC688765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109512.1 | NP_001102982.1 | 1 | 562 | ||
XM_006253209.2 | XP_006253271.1 | 1 | 905 |