siRNA Details
Gene Symbol: B3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109492.1 | NP_001102962.1 | 2 | 1298 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
