siRNA Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109411.1 | NP_001102881.2 | 5 | 446 | ||
| XM_008771367.1 | XP_008769589.1 | 5 | 924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc1b| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109411.1 | NP_001102881.2 | 5 | 446 | ||
| XM_008771367.1 | XP_008769589.1 | 5 | 924 | |||