siRNA Details
| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768533.1 | XP_008766755.1 | 23 | 4315 | ||
| XM_008768534.1 | XP_008766756.1 | 22 | 4288 | |||
| XM_008768535.1 | XP_008766757.1 | 23 | 4150 | |||
| XM_008768536.1 | XP_008766758.1 | 22 | 4123 | |||

Robo1
Genomic Map