siRNA Details
| Gene Symbol: Cmya5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231789.2 | XP_006231851.1 | 2 | 673 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmya5| Gene Symbol: Cmya5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231789.2 | XP_006231851.1 | 2 | 673 | ||