siRNA Details
| Gene Symbol: LOC685716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001064969.5 | XP_001064969.4 | 4 | 1123 | ||
| XM_008768747.1 | XP_008766969.1 | 4 | 757 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC685716| Gene Symbol: LOC685716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001064969.5 | XP_001064969.4 | 4 | 1123 | ||
| XM_008768747.1 | XP_008766969.1 | 4 | 757 | |||