siRNA Details
| Gene Symbol: Lbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129880.2 | NP_001123352.1 | 3 | 382 | ||
| XM_008764533.1 | XP_008762755.1 | 5 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lbh| Gene Symbol: Lbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129880.2 | NP_001123352.1 | 3 | 382 | ||
| XM_008764533.1 | XP_008762755.1 | 5 | 531 | |||