siRNA Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044282.1 | NP_001037747.1 | 2 | 279 | ||
| XM_006246126.2 | XP_006246188.1 | 2 | 429 | |||
| GenBank mRNA | BC085899.1 | - | 2 | 279 | 7193416 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mat2b| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044282.1 | NP_001037747.1 | 2 | 279 | ||
| XM_006246126.2 | XP_006246188.1 | 2 | 429 | |||
| GenBank mRNA | BC085899.1 | - | 2 | 279 | 7193416 | |