siRNA Details
| Gene Symbol: Rnf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134986.1 | NP_001128458.1 | 4 | 933 | ||
| XM_008760714.1 | XP_008758936.1 | 4 | 2507 | |||
| GenBank mRNA | BC168657.1 | - | 4 | 933 | 7313587 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf180| Gene Symbol: Rnf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134986.1 | NP_001128458.1 | 4 | 933 | ||
| XM_008760714.1 | XP_008758936.1 | 4 | 2507 | |||
| GenBank mRNA | BC168657.1 | - | 4 | 933 | 7313587 | |